Learn More
Selleck Chemical LLC Melibiose S4768-25mg
SDP
Catalog No. 503109874
Encompass_Preferred
Melibiose (Galalpha1-6Glcbeta) is a nondigestible disaccharide that consists of one galactose and one glucose moiety in an alpha (1-6) glycosidic linkage
Catalog No. 50-310-9874 Supplier Selleck Chemical LLC Supplier No. S476825MG
Melibiose (Galalpha1-6Glcbeta) is a nondigestible disaccharide that consists of one galactose and one glucose moiety in an alpha (1-6) glycosidic linkage