Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melinamide | 14417-88-0 | 99.9% | 383.61 g/mol | C26H41NO | 100 MG
SDP

Catalog No. 5000245707
Encompass_Preferred

Melinamide is an amide derivative of unsaturated long-chain fatty acids used in research to study lipid absorption and acyltransferase-related pathways. It is reported to inhibit cholesterol absorption with an IC50 of 20.9 μM and is provided as a high-purity analytical standard for biochemical assays, including DMSO solutions.

  • High purity analytical standard (99.9%).
  • Molecular formula C26H41NO; molecular weight 383.61 g/mol.
  • Reported inhibitor of cholesterol absorption (IC50 20.9 μM).
  • Soluble in DMSO; available as 10 mM solution.
  • Supplied in multiple pack sizes, including 100 mg.

Catalog No. 50-002-45707 Supplier Medchemexpress LLC Supplier No. HY101722100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Melinamide is an amide derivative of unsaturated long-chain fatty acids used in research to study lipid absorption and acyltransferase-related pathways. It is reported to inhibit cholesterol absorption with an IC50 of 20.9 μM and is provided as a high-purity analytical standard for biochemical assays, including DMSO solutions.

  • High purity analytical standard (99.9%).
  • Molecular formula C26H41NO; molecular weight 383.61 g/mol.
  • Reported inhibitor of cholesterol absorption (IC50 20.9 μM).
  • Soluble in DMSO; available as 10 mM solution.
  • Supplied in multiple pack sizes, including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.