Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | >98.0% | 327.89 | C21H26ClN | 200 MG
SDP

Catalog No. 5000242778
Encompass_Preferred

Melitracen hydrochloride is a research-grade tricyclic antidepressant hydrochloride salt supplied for laboratory use. It is used in pharmacological studies of monoamine neurotransmitter uptake and serotonergic (5-HT) receptor pathways. Provided as a solid for in vitro and preclinical research; not for human or veterinary use.

  • Tricyclic antidepressant hydrochloride salt.
  • Molecular formula C21H26ClN.
  • Molecular weight 327.89.
  • Purity >98.0% (HPLC).
  • Available as a solid powder, commonly in 200 MG quantities.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-42778 Supplier Medchemexpress LLC Supplier No. HY108256200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Melitracen hydrochloride is a research-grade tricyclic antidepressant hydrochloride salt supplied for laboratory use. It is used in pharmacological studies of monoamine neurotransmitter uptake and serotonergic (5-HT) receptor pathways. Provided as a solid for in vitro and preclinical research; not for human or veterinary use.

  • Tricyclic antidepressant hydrochloride salt.
  • Molecular formula C21H26ClN.
  • Molecular weight 327.89.
  • Purity >98.0% (HPLC).
  • Available as a solid powder, commonly in 200 MG quantities.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.