Learn More
Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | 99.8% | 327.89 | C21H26ClN | 10 MG
SDP
Catalog No. 5000242669
Encompass_Preferred
Melitracen hydrochloride is a tricyclic antidepressant research compound supplied as the hydrochloride salt and used as a chemically characterized reference for laboratory studies. It inhibits norepinephrine and serotonin uptake and is provided in small, labeled packs for analytical and formulation work.
  • High chemical purity (reported ~99.8%) for analytical applications.
  • Hydrochloride salt form for consistent physicochemical properties.
  • Molecular weight 327.89 and molecular formula C21H26ClN.
  • Available in multiple small pack sizes suitable for research and testing.
  • Supplied with product documentation and batch information.
Catalog No. 50-002-42669 Supplier Medchemexpress LLC Supplier No. HY10825610MG
Melitracen hydrochloride is a tricyclic antidepressant research compound supplied as the hydrochloride salt and used as a chemically characterized reference for laboratory studies. It inhibits norepinephrine and serotonin uptake and is provided in small, labeled packs for analytical and formulation work.
  • High chemical purity (reported ~99.8%) for analytical applications.
  • Hydrochloride salt form for consistent physicochemical properties.
  • Molecular weight 327.89 and molecular formula C21H26ClN.
  • Available in multiple small pack sizes suitable for research and testing.
  • Supplied with product documentation and batch information.