Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | 99.8% | 327.89 g/mol | C21H26ClN | 1 ML
SDP

Catalog No. 5000243427
Encompass_Preferred

Melitracen hydrochloride is a research-grade small molecule antidepressant and anxiolytic agent supplied for in vitro pharmacology, mechanism-of-action studies, and analytical reference. The material is provided with solubility and stock-solution guidance, and recommended storage conditions for both solid and solution forms.

  • High purity of 99.8%.
  • CAS number 10563-70-9, formula C21H26ClN, molecular weight 327.89 g/mol.
  • Available as solid samples and as a pre-made 10 mM solution (1 mL) in DMSO.
  • Soluble in DMSO and water at up to 100 mg/mL; ultrasonic assistance may be required.
  • Storage: solid at 4°C; in solution store at -80°C up to 6 months or -20°C up to 1 month.
  • Includes stock solution preparation guidance for common concentrations.

Catalog No. 50-002-43427 Supplier Medchemexpress LLC Supplier No. HY10825610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Melitracen hydrochloride is a research-grade small molecule antidepressant and anxiolytic agent supplied for in vitro pharmacology, mechanism-of-action studies, and analytical reference. The material is provided with solubility and stock-solution guidance, and recommended storage conditions for both solid and solution forms.

  • High purity of 99.8%.
  • CAS number 10563-70-9, formula C21H26ClN, molecular weight 327.89 g/mol.
  • Available as solid samples and as a pre-made 10 mM solution (1 mL) in DMSO.
  • Soluble in DMSO and water at up to 100 mg/mL; ultrasonic assistance may be required.
  • Storage: solid at 4°C; in solution store at -80°C up to 6 months or -20°C up to 1 month.
  • Includes stock solution preparation guidance for common concentrations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.