Learn More
Medchemexpress LLC Melitracen hydrochloride | 10563-70-9 | MFCD00242905 | 99.8% | 327.89 g/mol | C21H26ClN | 50 MG
SDP
Catalog No. 5000242559
Encompass_Preferred
Melitracen hydrochloride is the hydrochloride salt of melitracen, supplied as a high-purity research standard for analytical, pharmacological, and formulation studies. It is a white to off-white solid (CAS 10563-70-9) with defined solubility and storage recommendations to support reproducible results.
  • High purity suitable for analytical and pharmacological research.
  • White to off-white solid with consistent physical properties.
  • Soluble in water and DMSO; may require sonication.
  • Store solid at 4°C, sealed and protected from moisture; solutions at -80°C (long term) or -20°C (short term).
  • Supplied in small pack sizes convenient for assay and preclinical work.
Catalog No. 50-002-42559 Supplier Medchemexpress LLC Supplier No. HY10825650MG
Melitracen hydrochloride is the hydrochloride salt of melitracen, supplied as a high-purity research standard for analytical, pharmacological, and formulation studies. It is a white to off-white solid (CAS 10563-70-9) with defined solubility and storage recommendations to support reproducible results.
  • High purity suitable for analytical and pharmacological research.
  • White to off-white solid with consistent physical properties.
  • Soluble in water and DMSO; may require sonication.
  • Store solid at 4°C, sealed and protected from moisture; solutions at -80°C (long term) or -20°C (short term).
  • Supplied in small pack sizes convenient for assay and preclinical work.