Learn More
Cayman Chemical Melitracen (hydrochloride), Purity: ≥98%, CAS: 10563-70-9, 50 mg
SDP
Catalog No. 502942229
Encompass_Preferred
A thymoleptic and tricyclic antidepressant; inhibits 5-HT uptake in rat brain synaptosomes (IC50 = 670 nM); increases the lethality of yohimbine in mice at 20 and 40 mg/kg
Catalog No. 50-294-2229 Supplier Cayman Chemical Supplier No. 4011750 MG
A thymoleptic and tricyclic antidepressant; inhibits 5-HT uptake in rat brain synaptosomes (IC50 = 670 nM); increases the lethality of yohimbine in mice at 20 and 40 mg/kg