Learn More
Bachem Melittin Trifluoroacetate
SDP
Supplier:  Bachem 4030808.0001
Encompass_Preferred
Melittin Trifluoroacetate, H-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH2, 20449-79-0, C131H229N39O31, Mr 2846.50, 1mg. Synthetic melittin, the main component of the honeybee venom. The amphiphilic nature of this antimicrobial peptide makes it a excellent model compound for monitoring lipid-protein interactions in membranes. Adade et al. observed that melittin peptide could kill Trypanosoma cruzi parasites by inducing different cell death pathways.
Catalog No. 50-259-832