Learn More
Enzo Life Sciences Melittin (natural) (5mg). CAS: 20449-79-0
SDP
Supplier:  Enzo Life Sciences ALX162006M005
Encompass_Preferred
Causes disruption of normal cellular activity and cell lysis. Binds calmodulin in a calcium-dependent manner. Activates phospholipase A2 (PLA2) and inhibits protein kinase C (PKC) by binding to the catalytic domain in a Mg-ATP sensitive manner. Antirheumatic. Formula: C131H229N39O31. MW: 2846.5. Purity: ≥85% (HPLC). Appearance: White to faint tan powder. MI: 14: 5824. Solubility: Soluble in PBS (5mg/ml) or water (5mg/ml). Long Term Storage: -20°C. Handling: Very hygroscopic. Protect from light.
Catalog No. 50-200-7833