Learn More
Genscript Corporation MELITTIN PEPTIDE
Supplier:  Genscript Corporation RP102900.5
Encompass_Preferred
This 26-residue polypeptide isolated from bee venom binds calmodulin in a Ca 2+ -dependent manner, activates phospholipase A2, and inhibits protein kinase C (IC50=5-7 mM) by binding to the catalytic domain in a Mg-ATP-sensitive manner.
Catalog No. 50-274-784