Learn More
Enzo Life Sciences Melittin (synthetic) (0.5mg). CAS: 37231-28-0
SDP
Supplier:  Enzo Life Sciences ALX162007MC05
Encompass_Preferred
PLA2 stimulatory bee venom peptide. Sequence is similar to Phospholipase A2 Activating Peptide (ALX-165-007). Activates PLA2 (5-fold at 10µg/ml) in BC3H1 cells. For additional technical information see melittin (natural) (ALX-162-006). Formula: C131H229N39O31. MW: 2846.5. Purity: ≥97% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7835