Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melittoside | 19467-03-9 | 524.47 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYN091510MM/1ML

Encompass_Preferred

Melittoside is a natural compound, supplied as a solution in DMSO. This product is intended for research use only.

  • Purity: 99.01%
  • Molecular formula: C21H32O15
  • Appearance: Solid, white to light yellow
  • Solubility: Soluble in DMSO at ≥ 50 mg/mL (95.33 mM)
  • Storage: -20°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month, protect from light

Catalog No. 50-004-15988


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.