Learn More
Medchemexpress LLC Melk-8a hydrochloride | 2096992-20-8 | 99.3% | 469.02 | C25H33ClN6O | 5 MG
SDP
Catalog No. 5000214288
Encompass_Preferred
MELK-8a hydrochloride is a potent research-use small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with an IC50 of 2 nM. It is supplied as a light yellow to yellow solid with high purity, and is intended for in vitro research applications only. Chemical properties include formula C25H33ClN6O and molecular weight 469.02. Recommended storage is 4°C sealed; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-14288 Supplier Medchemexpress LLC Supplier No. HY100368A5MG
MELK-8a hydrochloride is a potent research-use small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with an IC50 of 2 nM. It is supplied as a light yellow to yellow solid with high purity, and is intended for in vitro research applications only. Chemical properties include formula C25H33ClN6O and molecular weight 469.02. Recommended storage is 4°C sealed; in solvent store at -80°C for long-term stability.