Learn More
Medchemexpress LLC Melk-8a (hydrochloride) | 2096992-20-8 | 99.3% | 469.02 | C25H33ClN6O | 500 MG
SDP
Catalog No. 5000214336
Encompass_Preferred
MELK-8a hydrochloride is a potent, selective inhibitor of maternal embryonic leucine zipper kinase (MELK) with an enzymatic IC50 of 2 nM. Supplied as the hydrochloride salt, the compound shows cellular activity in cancer cell lines, with reported good solubility and permeability, and is intended for research use only.
  • Potent MELK inhibition (IC50 2 nM).
  • High purity (>99%).
  • Good solubility and permeability in cellular assays.
  • Demonstrated activity in cancer cell lines (MDA-MB-468, MCF-7).
  • Documentation available: product data sheet and safety data sheet.
Catalog No. 50-002-14336 Supplier Medchemexpress LLC Supplier No. HY100368A500MG
MELK-8a hydrochloride is a potent, selective inhibitor of maternal embryonic leucine zipper kinase (MELK) with an enzymatic IC50 of 2 nM. Supplied as the hydrochloride salt, the compound shows cellular activity in cancer cell lines, with reported good solubility and permeability, and is intended for research use only.
  • Potent MELK inhibition (IC50 2 nM).
  • High purity (>99%).
  • Good solubility and permeability in cellular assays.
  • Demonstrated activity in cancer cell lines (MDA-MB-468, MCF-7).
  • Documentation available: product data sheet and safety data sheet.