Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Melk-8a hydrochloride | 2096992-20-8 | 99.3% | 469.02 g·mol⁻¹ | C25H33ClN6O | 1 ML
SDP

Catalog No. 5000214300
Encompass_Preferred

MELK-8a hydrochloride is a small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with an in vitro IC50 of 2 nM. It is provided for research use in solution and solid formats and is used in biochemical and cellular studies to probe MELK-associated signaling pathways.

  • Potent MELK inhibitor with an IC50 of 2 nM.
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solids (5-500 mg).
  • High purity (≈99.3%) suitable for research applications.
  • Molecular weight 469.02 g·mol⁻¹.
  • CAS number 2096992-20-8 for unambiguous identification.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-14300 Supplier Medchemexpress LLC Supplier No. HY100368A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MELK-8a hydrochloride is a small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with an in vitro IC50 of 2 nM. It is provided for research use in solution and solid formats and is used in biochemical and cellular studies to probe MELK-associated signaling pathways.

  • Potent MELK inhibitor with an IC50 of 2 nM.
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solids (5-500 mg).
  • High purity (≈99.3%) suitable for research applications.
  • Molecular weight 469.02 g·mol⁻¹.
  • CAS number 2096992-20-8 for unambiguous identification.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.