Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MELK-8a hydrochloride | 2096992-20-8 | 99.3% | 469.02 g/mol | C25H33ClN6O | 25 MG
SDP

Catalog No. 5000214258
Encompass_Preferred

MELK-8a hydrochloride is a potent small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with a reported IC50 of 2 nM. Supplied as the hydrochloride salt with high chemical purity, it is intended for research use in biochemical and cellular studies targeting MELK-dependent pathways.

  • Potent MELK inhibition with an IC50 of 2 nM.
  • Hydrochloride salt for improved stability and handling.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 469.02 g/mol and formula C25H33ClN6O.
  • Soluble in DMSO; solution formats available for convenient dosing.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-14258 Supplier Medchemexpress LLC Supplier No. HY100368A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MELK-8a hydrochloride is a potent small-molecule inhibitor of maternal embryonic leucine zipper kinase (MELK) with a reported IC50 of 2 nM. Supplied as the hydrochloride salt with high chemical purity, it is intended for research use in biochemical and cellular studies targeting MELK-dependent pathways.

  • Potent MELK inhibition with an IC50 of 2 nM.
  • Hydrochloride salt for improved stability and handling.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 469.02 g/mol and formula C25H33ClN6O.
  • Soluble in DMSO; solution formats available for convenient dosing.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.