Learn More
Cayman Chemical MELK-8ahydrochlorIde 50mg
SDP
Catalog No. 502915516
Encompass_Preferred
A MELK inhibitor (IC50 = 0.002 µM); selective for MELK over FLT3, haspin, KIT, and PDGFRα, (IC50s = 0.18, 0.19, >10, and 0.42 µM, respectively), as well as a panel of 456 kinases at 1 µM; inhibits the proliferation of MDA-MB-468 and MCF-7 cells (IC50s = 0.11 and 3.68 µM)
Catalog No. 50-291-5516 Supplier Cayman Chemical Supplier No. 3702550 MG
A MELK inhibitor (IC50 = 0.002 µM); selective for MELK over FLT3, haspin, KIT, and PDGFRα, (IC50s = 0.18, 0.19, >10, and 0.42 µM, respectively), as well as a panel of 456 kinases at 1 µM; inhibits the proliferation of MDA-MB-468 and MCF-7 cells (IC50s = 0.11 and 3.68 µM)