Learn More
Eurofins Discoverx MELK, active, 250 µg. (14-909M)
Supplier:  Eurofins Discoverx 14909M
Encompass
Recombinant human MELK (amino acids 1-340), containing an N-terminal GST tag. Maternal embryonic leucine zipper kinase (MELK) is a conserved serine/threonine kinase involved in a variety of cellular functions, including proliferation, differentiation and apoptosis. Its levels are elevated in a number of tumor types and it may be a prognosis marker for some cancers. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9398