Learn More
Moldex™ Mellows™ Plugstation™ Ear plug Dispenser
SDP
Catalog No. 19384344 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Holds:
250 Pairs
500 Pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19-384-344 500 Pairs
19-384-343 250 Pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-384-344 Supplier Moldex™ Supplier No. 6847
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mellows™ PlugStation™ dispensers are pre-filled with ear plugs and are convenient for any work environment. Easy access, wall-mountable unit will help ensure compliance. When the PlugStation is empty, simply dispose of it and hang a new one.
  • Holds Mellows™ disposable ear plugs
  • PlugStations™ are sealed to ensure ear plugs stay clean and sanitary
  • Highly visible to increase compliance
  • Dispenses one pair at a time to eliminate wasteful grabbing
  • Includes mounting bracket
  • Available in two sizes - small 250 Pair/Dispenser or large 500 Pair/Dispenser
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
High Visibility Yes
Holds 500 Pairs
Pairs per Package 500 Pairs/Dispenser
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 30 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Material PVC-free Plastic
Mounting Wall Mountable
Includes Mounting Hardware