Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM1701P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: M2-7C10 + M2-9E3 + HMB45 + T311. Clonality: Monoclonal. Host: Mouse. Isotype: IIIgG2b kappa. Positive Control: SK-MEL-13, SK-MEL-19 Melanoma cell lines; Melanomas. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20-22kDa (doublet) (MART); 70-80kDa (Tyrosinase); 90-100kDa (gp100).Chromosome: Multiple. Gene ID: 6490. Ensemble: ENSG00000185664. Uniprot: NP_001307051.
Catalog No. 50-188-5425