Learn More
Cayman Chemical MelnocortIn 2MC2RACTHR 96Wel
SDP
Catalog No. 502945444
Encompass_Preferred
Human MC2R Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB41001]
Catalog No. 50-294-5444 Supplier Cayman Chemical Supplier No. 4255696 WELL
Human MC2R Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. IB41001]