Learn More
Cayman Chemical Melanocortin-4 Receptor Reporter Assay Kit, 1 ea
SDP
Catalog No. 502894795
Encompass_Preferred
Cayman’s MC4R Reporter Assay consist of a 96-well plate coated with a transfection complex containing DNA constructs for MC4R and a cAMP response element regulated Secreted Alkaline Phosphatase (SEAP) reporter (MC4R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express MC4R at the cell surface. Binding of agonists to MC4R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture media. SEAP activity is measured following addition of a luminescence-based alkaline phosphatase substrate provided in the kit.
Catalog No. 50-289-4795 Supplier Cayman Chemical Supplier No. 6001901 EA
Cayman’s MC4R Reporter Assay consist of a 96-well plate coated with a transfection complex containing DNA constructs for MC4R and a cAMP response element regulated Secreted Alkaline Phosphatase (SEAP) reporter (MC4R Reverse Transfection Strip Plate). Cells grown on the transfection complex will express MC4R at the cell surface. Binding of agonists to MC4R initiates a signal transduction cascade resulting in expression of SEAP which is secreted into the cell culture media. SEAP activity is measured following addition of a luminescence-based alkaline phosphatase substrate provided in the kit.