Learn More
Cayman Chemical MelnostatInfrogeeltrI 10mg
SDP
Catalog No. 502944152
Encompass_Preferred
An endogenous peptide; inhibits α-MSH release from primary frog NILs (ED50 = 100 nM); inhibits thyrotropin-releasing hormone-induced increases in intracellular calcium in primary frog melanotrope cells at 100 nM; increases food intake in frog larvae at 5 or 10 nmol/kg, i.c.v.; increases mean blood pressure in normotensive sharks at 5 nmol/kg
Catalog No. 50-294-4152 Supplier Cayman Chemical Supplier No. 4146110 MG
An endogenous peptide; inhibits α-MSH release from primary frog NILs (ED50 = 100 nM); inhibits thyrotropin-releasing hormone-induced increases in intracellular calcium in primary frog melanotrope cells at 100 nM; increases food intake in frog larvae at 5 or 10 nmol/kg, i.c.v.; increases mean blood pressure in normotensive sharks at 5 nmol/kg