Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Melon™ Gel Chromatography Cartridges

Catalog No. p-4527403 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 cartridge x 5 mL
2 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89932 2 x 1 mL
PI89933 1 cartridge x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI89932 Supplier Thermo Scientific™ Supplier No. 89932
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly purify human, mouse and rabbit IgG from serum and other sources with this simpler alternative to traditional Protein A and Protein G methods.

Thermo Scientific™ Melon Gel IgG Purification Resins and Kits comprise a simpler and faster alternative to traditional Protein A and Protein G methods for purification of human, mouse and rabbit IgG from serum and other sources.

Melon Gel IgG Purification Kits enable fast, easy and gentle purification of antibodies from serum and other sample types. Antibody purification with Melon Gel may be performed with gravity-flow columns, spin columns or chromatography cartridges. Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.

Highlights:

  • Simple, one-step protocol – IgG Purification with Melon Gel Resin does not require the binding, washing, and multiple elution steps required by Protein A and Protein G

  • Rapid purification – antibody purification from serum is four to six times faster than Protein A or G methods

  • High recovery and purity – antibody recovery from serum for many species is greater than 90% with greater than 80% purity

  • Minimal dilution – reduced need to concentrate antibody before storage or use

  • Robust purification – Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G

  • Gentle purification – no harsh antibody elution conditions helps retain IgG activity

  • Reusable support – Melon Gel Resin can be used for multiple antibody purifications

  • Available in various formats – spin columns, purification kits and chromatography cartridges

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chromatography Cartridge
Product Line Melon
Column Type Proprietary Resin, Affinity
Format Chromatography Cartridge
Stationary Phase Proprietary Ligand
Flow Rate 2 mL/min.
Purification Target IgG Antibodies
Volume (Metric) Bed 1 mL
Content And Storage Upon receipt store at 4°C to 8°C. Do not freeze.
Quantity 2 x 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.