Learn More
Medchemexpress LLC HY-B0261 100mg Medchemexpress, Meloxicam CAS:71125-38-7 Purity:>98%
SDP
Catalog No. 502082431
Encompass_Preferred
Medchemexpress, HY-B0261 100mg Meloxicam CAS:71125-38-7 Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 µM and 36.6 µM for COX-2 and COX-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2431 Supplier Medchemexpress LLC Supplier No. HYB0261100MG
Medchemexpress, HY-B0261 100mg Meloxicam CAS:71125-38-7 Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 µM and 36.6 µM for COX-2 and COX-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.