Learn More
Medchemexpress LLC Meloxicam-13C,d3 | 1309936-00-2 | ≥99.9% | 1 MG
SDP
Catalog No. 5000407381
Encompass_Preferred
Meloxicam-13C,d3 is a deuterium labeled non-steroidal anti-inflammatory agent that inhibits COX activity, with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1. It is for research use only and not sold to patients. Stable heavy isotopes have been incorporated into this drug molecule, primarily as a tracer for quantitation during drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07381 Supplier Medchemexpress LLC Supplier No. HYB0261S21MG
Meloxicam-13C,d3 is a deuterium labeled non-steroidal anti-inflammatory agent that inhibits COX activity, with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1. It is for research use only and not sold to patients. Stable heavy isotopes have been incorporated into this drug molecule, primarily as a tracer for quantitation during drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs