Learn More
Medchemexpress LLC Meloxicam-d3-1 | 1227358-55-5 | 98.0% | 1 MG
SDP
Catalog No. 5000407380
Encompass_Preferred
Meloxicam-d3-1 is a deuterium-labeled Meloxicam, which acts as a non-steroidal anti-inflammatory agent. It inhibits COX activity, with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07380 Supplier Medchemexpress LLC Supplier No. HYB0261S11MG
Meloxicam-d3-1 is a deuterium-labeled Meloxicam, which acts as a non-steroidal anti-inflammatory agent. It inhibits COX activity, with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.