Learn More
Med Vet International Meloxicam Injection (metacam), 5mg/mL, 10mL
Catalog No. 503045377
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dogs Meloxicam Solution for Injection is indicated in dogs for the control of pain and inflammation associated with osteoarthritis. Cats For the control of postoperative pain and inflammation associated with orthopedic surgery, ovariohysterectomy and castration when administered prior to surgery.
Catalog No. 50-304-5377 Supplier Med Vet International Supplier No. RXMELOXICAMINJ
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dogs Meloxicam Solution for Injection is indicated in dogs for the control of pain and inflammation associated with osteoarthritis. Cats For the control of postoperative pain and inflammation associated with orthopedic surgery, ovariohysterectomy and castration when administered prior to surgery.