Learn More
Apexbio Technology LLC Meloxicam (Mobic) 71125-38-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061844
Encompass_Preferred
Meloxicam (CAS 71125-38-7) commercially known as Mobic is a nonsteroidal anti-inflammatory drug (NSAID) that selectively inhibits cyclooxygenase-2 (COX-2) thereby interfering with prostaglandin synthesis and mitigating inflammation and pain Its pharmacological activity against COX-2 reduces symptoms associated with rheumatoid arthritis juvenile rheumatoid arthritis and osteoarthritis such as joint pain stiffness and swelling As a COX-2 preferential inhibitor meloxicam is widely utilized as a reference compound in biomedical research examining inflammatory signaling pathways and arthritis-related disease models
Catalog No. 50-000-61844 Supplier Apexbio Technology LLC Supplier No. A84665.1
Meloxicam (CAS 71125-38-7) commercially known as Mobic is a nonsteroidal anti-inflammatory drug (NSAID) that selectively inhibits cyclooxygenase-2 (COX-2) thereby interfering with prostaglandin synthesis and mitigating inflammation and pain Its pharmacological activity against COX-2 reduces symptoms associated with rheumatoid arthritis juvenile rheumatoid arthritis and osteoarthritis such as joint pain stiffness and swelling As a COX-2 preferential inhibitor meloxicam is widely utilized as a reference compound in biomedical research examining inflammatory signaling pathways and arthritis-related disease models