Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meloxicam (Standard) | 71125-38-7 | 99.89% | 25 MG
SDP

Catalog No. 5000450563
Encompass_Preferred

Meloxicam (Standard) is the analytical standard of Meloxicam, a non-steroidal anti-inflammatory agent that inhibits COX activity. It is intended for research and analytical applications, commonly used as a reference standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Inhibits COX activity with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.
  • Has a molecular weight of 351.40.
  • Chemical formula is C14H13N3O4S2.
  • Appearance is a solid, light yellow to green yellow in color.
  • Purity is 99.89%.

Catalog No. 50-004-50563 Supplier Medchemexpress LLC Supplier No. 25MGHYB0261R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meloxicam (Standard) is the analytical standard of Meloxicam, a non-steroidal anti-inflammatory agent that inhibits COX activity. It is intended for research and analytical applications, commonly used as a reference standard in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Inhibits COX activity with IC50s of 0.49 μM for COX-2 and 36.6 μM for COX-1.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.
  • Has a molecular weight of 351.40.
  • Chemical formula is C14H13N3O4S2.
  • Appearance is a solid, light yellow to green yellow in color.
  • Purity is 99.89%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.