Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meloxicam, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604289 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-289 5 g Amber Glass Bottle
18-604-290 25 g Amber Glass Bottle
18-604-291 100 g Amber Glass Bottle
18-604-292 500 g Amber Glass Bottle
18-604-293 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-289 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M17785GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

M1778, 71125-38-7, C14H13N3O4S2

Spectrum™ Chemical Meloxicam, USP is a nonsteroidal anti-inflammatory used to treat arthritis. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
IUPAC Name (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ6,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Meloxicam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.