Learn More
Cayman Chemical Melperone-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1219798-80-7, 5 mg
SDP
Catalog No. 502991921
Encompass_Preferred
An internal standard for the quantification of melperone by GC- or LC-MS
Catalog No. 50-299-1921 Supplier Cayman Chemical Supplier No. 314895 MG
An internal standard for the quantification of melperone by GC- or LC-MS