Learn More
Medchemexpress LLC Melperone-d4 hydrochloride | 1219798-80-7 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB2169S21MG
Encompass_Preferred
Melperone-d4 hydrochloride is a deuterium-labeled form of Melperone hydrochloride, a butyrophenone with atypical antipsychotic properties. It functions as a multireceptor antagonist, targeting 5-HT2A, dopamine D2, α1-adrenergic, and α2-adrenergic receptors, along with weaker binding to other receptors. It also inhibits CYP2D6. This compound is valuable for research into schizophrenia and agitation in the elderly.
  • Deuterium-labeled compound
  • Atypical antipsychotic properties
  • Functions as a multireceptor antagonist
  • Targets 5-HT2A, dopamine D2, α1-adrenergic, and α2-adrenergic receptors
  • Inhibits CYP2D6
  • Useful for research on schizophrenia and agitation in the elderly
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-10331