Learn More
M+A Matting Melt Step™ Cord with GFCI
Catalog No. 17555117 Shop All M+A Matting Products
Change view
Click to view available options
Length (English):
6 ft.
Catalog No. Length (English)
17-555-117 6 ft.
1 options
Catalog No. 17-555-117 Supplier M+A Matting Supplier No. 889000006
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Melt Step™ Cord with GFCI works in conjuction with Melt Step™ Matting to keep walkways free of snow and ice to minimize the risk of slip-and-fall accidents.
  • Allows mat to heat to approximately 40 to 45˚F above ambient air temperature
  • Waterproof electrical connection system with GFCI switch prevents electrical shock hazard
  • Up to 4 mats can be connected in sequence to cover up to 20 linear feet of walkway
Specifications
Description Accessory for Melt Step: Cord + GFCI
Length (English) 6 ft.