Learn More
Description
- Effective – Mat will heat to approximately 40 to 45˚F above ambient air temperature
- Safe – Waterproof electrical connection system with GFCI switch prevents electrical shock hazard
- Certified high-traction by the National Floor Safety Institute (NFSI)
- Durable – Molded nitrile rubber construction is waterproof, UV-resistant, and flexible even in freezing temperatures
- Scalable – Up to 4 mats can be connected in sequence to cover up to 20 linear feet of walkway
- All Melt Step mats are 3 x 5 ft. with choice of one connector or two connectors; unique reversible design of the mat permits electrical connection on the right or left side of the walkway
- 6 ft. power cord (120 volts) with GFCI switch as well as 6 ft. extension cord are both sold separately
Specifications
Specifications
| Dimensions (L x W) | 3 x 5 ft. |
| Includes | 1 connection |
| Certifications/Compliance | NFSI Certified. Made in USA |
| Color | Black |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.