Learn More
M+A Matting Melt Step™ Waterproof Extension Cord
Catalog No. 17555115 Shop All M+A Matting Products
Change view
Click to view available options
Length (English):
6 ft.
Catalog No. Length (English)
17-555-115 6 ft.
1 options
Catalog No. 17-555-115 Supplier M+A Matting Supplier No. 889010006
Melt Step™ Waterproof Extension Cord works in conjuction with Melt Step™ Matting to keep walkways free of snow and ice to minimize the risk of slip-and-fall accidents.
  • Allows mat to heat to approximately 40 to 45˚F above ambient air temperature
  • Waterproof electrical connection system prevents electrical shock hazard
  • Up to 4 mats can be connected in sequence to cover up to 20 linear feet of walkway
Specifications
Description Accessory for Melt Step: Extension cord
Length (English) 6 ft.