Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melting point standard (182-184°C), MilliporeSigma™ Supelco™

Catalog No. 111012269 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2269 250 mg
11-101-2268 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2269 Supplier MilliporeSigma Supelco Supplier No. 42183250MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-09-4
Flash Point 185°C
Physical Form Neat
Linear Formula CH3 OC6H4 CO2 H
MDL Number MFCD00002542
Melting Point 182°C to 184°C (±0.3); 182°C to 185°C
Synonym p-Anisic acid; 4-Methoxybenzoic acid; p-Methoxybenzoic acid; Draconic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 152.15
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.