Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melting point Standard 283-286C, MilliporeSigma™ Supelco™

Catalog No. 111015674 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5674 250 mg
11-101-5673 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5674 Supplier MilliporeSigma Supelco Supplier No. 67372250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anthraquinone
CAS 84-65-1
Vapor Pressure 1 mmHg (190°C)
Vapor Density 7.16 (vs. air)
Flash Point 185°C - closed cup
Physical Form Neat
Linear Formula C14H8O2
MDL Number MFCD00001188
Melting Point 283°C to 286°C (±0.3 C); 284°C to 286°C
Boiling Point 379°C to 381°C
Synonym Anthraquinone
Type Analytical Standard
Industry Type Laboratory Use
Molecular Weight (g/mol) 208.21
Quantity 250 mg
Formula Weight 208.21
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.