Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melting Point Standard, 47-49°C, MilliporeSigma™ Supelco™

Catalog No. 111020667 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0667 1 g
11-101-9503 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0667 Supplier MilliporeSigma Supelco Supplier No. 502961G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 119-61-9
Vapor Pressure 1 mmHg (108°C)
Physical Form Neat
Linear Formula (C6H5)2 CO
MDL Number MFCD00003076
Melting Point 47°C to 49°C (±0.3 C); 47°C to 51°C
Boiling Point 305°C
Synonym Benzophenone; Diphenyl ketone; Diphenylmethanone; NSC 8077
Quantity 1 g
Formula Weight 182.22
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.