Learn More
Electron Microscopy Sciences Meltmount™ Slide Mounting Media Quick-Stick™ 1.539
SDP
Catalog No. 5027096
Encompass_Preferred
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) Used to make permanent microscope slide mounts quickly by can be appling to slide on a hotplate. Can be reversed by reheating Quick-Stick. Weighs approximately 2/3 oz. (18g).
Refractive index: 1.539
Catalog No. 50-270-96 Supplier Electron Microscopy Sciences Supplier No. 1799411
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) Used to make permanent microscope slide mounts quickly by can be appling to slide on a hotplate. Can be reversed by reheating Quick-Stick. Weighs approximately 2/3 oz. (18g).
Refractive index: 1.539