Learn More
Electron Microscopy Sciences Meltmount™ Slide Mounting Media Quick-Stick™ 1.662
SDP
Catalog No. 5027106
Encompass_Preferred
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) It has a refractive index (nD @25 deg C) of 1.662 and an Abbe V dispersion of 26, optically similar to Aroclor 5442 but is PCB-free.
Refractive index: 1.662
Catalog No. 50-271-06 Supplier Electron Microscopy Sciences Supplier No. 1799451
Cargille Meltmount (optical quality) is a series of mounting media which are specifically formulated for use in microscope slide mounting and in other optical coupling applications.
< Use Meltmount for instant thermal mountants that require "no-oven-time" and are thermally reversible for sample retrieval or re-orientation. Meltmounts are PBC free and contains no solvents. Cleanup in toluene. Fluid at 65 degree C. It is a direct replacement for old mountant mediums (Canada Balsam, Aroclor 5442, Naphrax...) It has a refractive index (nD @25 deg C) of 1.662 and an Abbe V dispersion of 26, optically similar to Aroclor 5442 but is PCB-free.
Refractive index: 1.662