Learn More
Cayman Chemical Melittoside, Purity: ≥98%, CAS: 19467-03-9, 10 mg
SDP
Catalog No. 502940576
Encompass_Preferred
An iridoid glycoside with antioxidant and enzyme inhibitory activities; scavenges free radicals in a TEAC assay; inhibits the activity of AChE and BChE by 24.6 and 12.8%, respectively, at 250 µg/ml in a cell-free assay
Catalog No. 50-294-0576 Supplier Cayman Chemical Supplier No. 2903510 MG
An iridoid glycoside with antioxidant and enzyme inhibitory activities; scavenges free radicals in a TEAC assay; inhibits the activity of AChE and BChE by 24.6 and 12.8%, respectively, at 250 µg/ml in a cell-free assay