Learn More
Electron Microscopy Sciences MEM-32 Grid Storage Box
SDP
Catalog No. 50930878
Encompass_Preferred
This multipurpose electron microscope specimen grid box eliminates the chances of tweezer insertion damaging the grids because the tweezers slot' and grid slot' are in a separate location. The tweezers are only able to grip the edge of the grid enabling it to be picked up. And the grids stored no longer jump out of the box while you remove the cover. Between the body of the box and the sliding lid, there is a separate plastic cover, which allows foronly four slots beingexposed at a time. The Grid Record Card is stored safely by insertion along on the reverse side of the box.
Catalog No. 50-930-878 Supplier Electron Microscopy Sciences Supplier No. 7116510
This multipurpose electron microscope specimen grid box eliminates the chances of tweezer insertion damaging the grids because the tweezers slot' and grid slot' are in a separate location. The tweezers are only able to grip the edge of the grid enabling it to be picked up. And the grids stored no longer jump out of the box while you remove the cover. Between the body of the box and the sliding lid, there is a separate plastic cover, which allows foronly four slots beingexposed at a time. The Grid Record Card is stored safely by insertion along on the reverse side of the box.