Learn More
ILEX LIFE SCIENCES LLC MEM EBSS P04-00509 500 ML
SDP
Catalog No. 502733906
Encompass_Preferred
PAN-Biotech MEM Eagle Cell Culture Medium w: EBSS, w: L-Glutamine, w: 1.5 g/L NaHCO3 (500 ml). EBSS: Contains Earle’s Balanced Salt Solution. BICARBONATE: With 1.5 g/L Sodium Bicarbonate GLUCOSE: With glucose. GLUTAMINE: With L-Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Minimum Essential Medium (MEM), developed by Harry Eagle, is a widely used synthetic cell culture media. After finding that Eagle's Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells, MEM was created with higher concentrations of amino acids to more closely resemble mammalian cell protein composition. When supplemented with serum, MEM is suitable for cultivation of a wide variety of cells grown in monolayers. MEM often serves as a base medium for many further modifications.
Catalog No. 50-273-3906 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0400509
Add to Cart
Add to Cart
PAN-Biotech MEM Eagle Cell Culture Medium w: EBSS, w: L-Glutamine, w: 1.5 g/L NaHCO3 (500 ml). EBSS: Contains Earle’s Balanced Salt Solution. BICARBONATE: With 1.5 g/L Sodium Bicarbonate GLUCOSE: With glucose. GLUTAMINE: With L-Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Minimum Essential Medium (MEM), developed by Harry Eagle, is a widely used synthetic cell culture media. After finding that Eagle's Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells, MEM was created with higher concentrations of amino acids to more closely resemble mammalian cell protein composition. When supplemented with serum, MEM is suitable for cultivation of a wide variety of cells grown in monolayers. MEM often serves as a base medium for many further modifications.