Learn More
ILEX LIFE SCIENCES LLC MEM EBSS P04-08056 6 X 500 ML
SDP
Catalog No. 502733857
Encompass_Preferred
PAN-Biotech MEM Eagle Cell Culture Medium w: EBSS, w: 2 mM L-Glutamine, w: 1 mM Sodium pyruvate, w: NEAA, w: 1.5 g/L NaHCO3 (6 x 500 ml). EBSS: Contains Earle’s Balanced Salt Solution. MEM-NEAA: With non-essential amino acids (NEAA) to MEM (MEM-NEAA). BICARBONATE: With 1.5 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With 2 mM L-Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: With 1 mM Sodium Pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Minimum Essential Medium (MEM), developed by Harry Eagle, is a widely used synthetic cell culture media. After finding that Eagle's Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells, MEM was created with higher concentrations of amino acids to more closely resemble mammalian cell protein composition.
Catalog No. 50-273-3857 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0408056X6
Add to Cart
Add to Cart
PAN-Biotech MEM Eagle Cell Culture Medium w: EBSS, w: 2 mM L-Glutamine, w: 1 mM Sodium pyruvate, w: NEAA, w: 1.5 g/L NaHCO3 (6 x 500 ml). EBSS: Contains Earle’s Balanced Salt Solution. MEM-NEAA: With non-essential amino acids (NEAA) to MEM (MEM-NEAA). BICARBONATE: With 1.5 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With 2 mM L-Glutamine. HEPES: Without HEPES. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: With 1 mM Sodium Pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Minimum Essential Medium (MEM), developed by Harry Eagle, is a widely used synthetic cell culture media. After finding that Eagle's Basal Medium (BME) did not meet specific nutritional requirements of normal mammalian fibroblasts and certain HeLa cells, MEM was created with higher concentrations of amino acids to more closely resemble mammalian cell protein composition.