Learn More
Tisch Scientific Tisch Scientific MCE Membrane Filter, 5.0 µm, 47 mm, Nonsterile, 200 Pack, SF14634
SDP
Supplier:  Tisch Scientific SF14634
Encompass
Tisch MCE membrane filters are developed from a mixture of cellulose acetate and cellulose nitrate and are one of the most widely used filters in both air monitoring and analytical applications.
Catalog No. NC1709849