Learn More
ATHENA ES MEM with L-Glutamine
Catalog No. 5000158125
Encompass_Preferred
Minimum Essential Media (MEM), patterned after Eagle’s Media, is one of the most commonly used of all cell culture media and is well suited for the growth of a broad spectrum of mammalian cells. Compared to the earlier Basal Medium Eagle (BME), MEM is formulated with increased concentrations of amino acids. Because MEM media lacks proteins, lipids, or growth factors, it commonly requires supplementation with fetal bovine serum (FBS).
Catalog No. 50-001-58125 Supplier ATHENA ES Supplier No. 0464
Minimum Essential Media (MEM), patterned after Eagle’s Media, is one of the most commonly used of all cell culture media and is well suited for the growth of a broad spectrum of mammalian cells. Compared to the earlier Basal Medium Eagle (BME), MEM is formulated with increased concentrations of amino acids. Because MEM media lacks proteins, lipids, or growth factors, it commonly requires supplementation with fetal bovine serum (FBS).