Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mem-PER™ Eukaryotic Membrane Protein Extraction Kit

Catalog No. PI89826 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
75 mL Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89826 75 mL Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89826 Supplier Thermo Scientific™ Supplier No. 89826
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Perform small-scale hydrophobic membrane-associated protein solubilization and enrichment with this simple reagent-based procedure.

Thermo Scientific™ Mem-PER Eukaryotic Membrane Protein Extraction Kit enables small-scale solubilization and enrichment of hydrophobic membrane-associated proteins in a simple reagent-based procedure.

Traditional methods for isolation of membrane protein are tedious and time-consuming, and they require gradient separation and expensive ultracentrifugation equipment. The Mem-PER Kit effectively isolates membrane proteins from cultured cells and tissues using a simple benchtop microcentrifuge procedure. Hydrophobic proteins having three or fewer membrane-spanning domains are effectively solubilized and isolated from hydrophilic (cytoplasmic) proteins using the three-reagent system. The kit makes possible the extraction and selective enrichment of integral and attached membrane proteins from cultured mammalian and yeast cells, as well as from hard and soft mammalian tissues.

Highlights:

  • Extraction and isolation – produces minimal cross-contamination (typically <10%) of hydrophilic protein into the hydrophobic (membrane) protein fraction

  • Cells or tissues – extracts protein from cultured mammalian and yeast cells and mammalian tissues

  • Downstream compatibility – analyze membrane protein extracts by SDS-PAGE, Western blotting and protein assays

  • Fast and simple – isolate membrane proteins in approximately one hour

  • No special equipment required – requires only a benchtop microcentrifuge, tubes and pipetters

Includes:

Three buffers and detergent solutions necessary to make working reagents

TRUSTED_SUSTAINABILITY

Specifications

Formulation Three-component kit
Description Mem-PER Eukaryotic Membrane Protein Extraction Kit
Includes Mem-PER Reagent C, 40mL; Mem-PER Reagent B, 25mL; Mem-PER Reagent A, 10mL
Target Proteins Membrane
Donor Source Cultured Mammalian Cells, Cultured Yeast Cells, Animal Tissue
Sufficient For 60 to 80 samples each containing 5 million cells, 15mg yeast cell paste or 20mg tissue
Quantity 75 mL Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.