Learn More
Molecular Dimensions MemTrans™
SDP
Catalog No. NC1604553
Encompass
A unique screen designed for transporter proteins. MemTrans uniquely targets transporter proteins. Includes the most common transporter protein crystallization conditions to maximize your chances of success.
Catalog No. NC1604553 Supplier Molecular Dimensions Supplier No. MD1113
A unique screen designed for transporter proteins. MemTrans uniquely targets transporter proteins. Includes the most common transporter protein crystallization conditions to maximize your chances of success.