Learn More
Sigma Aldrich Fine Chemicals Biosciences MEM Vitamin Solution 100x
Catalog No. 502901751
Encompass
Synthetic/ basal media used currently contain amino acids carbohydrates vitamins and salts. Eagle minimum essential medium (MEM) is a modified version of BME (basal medium Eagle) which contains increased amounts of each amino acid. Addition of water soluble vitamins such as inositol to various culture facilitates the hatching of hamster and rabbit blastocysts. Vitamins also influence the metabolism of glucose in sheep and mouse embryos.
Catalog No. 50-290-1751 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M6895100ML
Synthetic/ basal media used currently contain amino acids carbohydrates vitamins and salts. Eagle minimum essential medium (MEM) is a modified version of BME (basal medium Eagle) which contains increased amounts of each amino acid. Addition of water soluble vitamins such as inositol to various culture facilitates the hatching of hamster and rabbit blastocysts. Vitamins also influence the metabolism of glucose in sheep and mouse embryos.